missing translation for 'onlineSavingsMsg'
Learn More

C9orf11 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 23168-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892215

  • 3468.15 NOK / 150µL
Estimated Shipment: 13-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

C9orf11
Polyclonal
Unconjugated
EQTN
Afaf, C9orf11, EQTN, Equatorin
Rabbit
Antigen Affinity Chromatography
RUO
54586
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.65 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NQ60
EQTN
C9orf11 Fusion Protein Ag19656
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers