missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ C18orf43 Recombinant Protein

Brand:  Abnova™ H00010650-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135502

  • 4115.00 NOK / 10µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH07528
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
31.9
8
Glutathione Sepharose 4 Fast Flow
10 ug
MRGEGWGPGVSRPFLQAVPAPSSLPLPCPGDILRPGWRPCLAACQPHLLGLEAMDPS
C18orf43/DKFZp547K106/FLJ31484/HFL-EDDG1/MGC120170/PRELID3A
SLMO1
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
10650
C18orf43 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SLMO1
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers