missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C17orf46 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | C17orf46 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803125
|
Proteintech
24629-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16893115
|
Proteintech
24629-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
C17orf46, also named as SPATA32 and TEX34, mainly expressed in Testis. It interacts with syntaxin-1 and ACTB. It′s about 42-52 kDa in our detection.Specifications
C17orf46 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q96LK8 | |
124783 | |
C17orf46 Fusion Protein Ag18239 | |
Primary | |
-20°C | |
SPATA32 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C17orf46 | |
SPATA32 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |