missing translation for 'onlineSavingsMsg'
Learn More

C10orf46 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26321-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16884825

  • 1281.92 NOK / 20µL
Estimated Shipment: 16-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cell cycle associated protein capable of promoting cell proliferation through the activation of CDK2 at the G1/S phase transition.
Specifications

Specifications

C10orf46
Polyclonal
Unconjugated
CACUL1
C10orf46, CACUL1
Rabbit
Antigen Affinity Chromatography
RUO
143384, 78832
-20°C
Liquid
Western Blot
0.46 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86Y37, Q8R0X2
Cacul1
C10orf46 Fusion Protein Ag23694
20 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers