Learn More
C-Peptide Antibody (CC27) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP1-05434JF585
Product Code. 30171201
  • 4990.00 NOK / 0.20mL
Description
C-Peptide Monoclonal antibody specifically detects C-Peptide in Rat samples. It is validated for ELISA,Immunoassay,Sandwich ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C-Peptide
Monoclonal
Janelia Fluor 585
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of rat C-peptides I and II conjugated with the carrier protein.
0.2 mL
Diabetes Research, Lipid and Metabolism
3630
Store at 4°C in the dark.
IgG1
ELISA, Immunoassay, Sandwich ELISA
CC27
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Rat