Learn More
Uvex Busingess Gray PU 84697 Low Shoe - Width 10
Uvex 1 shoes designed for classic business look while providing protection against workplace risks; extra-wide steel toecap, penetration-resistant midsole and SRC polyurethane outsole protects toes and keeps from slipping; ideal for active men in office and factory environments
1435.00 NOK
Specifications
Color | Gray |
---|---|
Material | Microvelour |
Sole Material | Polyurethane (PU) |
Toe Material | Steel |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17214571
|
Uvex
8469739 |
39 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17224571
|
Uvex
8469740 |
40 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17234571
|
Uvex
8469741 |
41 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17244571
|
Uvex
8469742 |
42 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17254571
|
Uvex
8469743 |
43 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17264571
|
Uvex
8469744 |
44 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17274571
|
Uvex
8469745 |
45 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17284571
|
Uvex
8469746 |
46 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17294571
|
Uvex
8469747 |
47 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17204581
|
Uvex
8469748 |
48 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17214581
|
Uvex
8469749 |
49 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17224581
|
Uvex
8469750 |
50 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17234581
|
Uvex
8469751 |
51 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17244581
|
Uvex
8469752 |
52 |
1435.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
Description
Uvex 1 business Shoe 8469.7 S2 SRC
- Safe, smart, stylish
- Fashionably business look with trendy design
- Generous fit with wide steel toe cap
- Uvex Climazone technology ensures optimum wearer comfort
- Breathable, water-resistant high-tech microvelour upper material
- Textile lining provides optimum breathability
- Made from synthetic materials, suitable for people allergic to chrome
- Shock absorbing, durable mono density polyurethane sole unit
- Removable comfort insole, antistatic with moisture control system and shock absorption in heel and at front of foot
- Soft padded collar and tongue
- Sole unit of free of lacquer wetting inhibitors such as silicones, phthalates, etc.
- Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 8469.7
- Standard EN ISO 20345:2011 S2 SRC
- Sizes: 39 to 52
Specifications
Gray | |
Polyurethane (PU) | |
No | |
Men's | |
Uvex 1 | |
No | |
SRC |
Microvelour | |
Steel | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S2 |