Learn More
Uvex Busingess Blue PU 84263 Boot - Width 12
Fashionable business look with a trendy design includes steel toecap
1530.00 NOK
Specifications
Color | Blue |
---|---|
Material | Microvelour |
Sole Material | Polyurethane (PU) |
Toe Material | Steel |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17224471
|
Uvex
8426339 |
39 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17234471
|
Uvex
8426340 |
40 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17244471
|
Uvex
8426341 |
41 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17254471
|
Uvex
8426342 |
42 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17264471
|
Uvex
8426343 |
43 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17274471
|
Uvex
8426344 |
44 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17284471
|
Uvex
8426345 |
45 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17294471
|
Uvex
8426346 |
46 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17204481
|
Uvex
8426347 |
47 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17214481
|
Uvex
8426348 |
48 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17224481
|
Uvex
8426349 |
49 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17234481
|
Uvex
8426350 |
50 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17244481
|
Uvex
8426351 |
51 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17254481
|
Uvex
8426352 |
52 |
1530.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
Uvex 1 business Chelsea Boot 8426.3 S3 SRC
- Safe, smart, stylish
- Fashionably business look with trendy design
- Generous fit with wide steel toe cap
- Penetration-resistant, non-metallic midsole in accordance with latest safety standards, does not restrict flexibility
- Uvex Climazone technology ensures optimum wearer comfort
- Breathable, water-resistant high-tech microvelour upper material
- Textile lining provides optimum breathability
- Made from synthetic materials, suitable for people allergic to chrome
- Shock absorbing, durable mono density polyurethane sole unit
- Removable comfort insole, antistatic with moisture control system and shock absorption in heel and at front of foot
- Sole unit of free of lacquer wetting inhibitors such as silicones, phthalates, etc.
- Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 8426.3
- Standard EN ISO 20345:2011 S3 SRC
- Sizes: 39 to 52
Specifications
Blue | |
Polyurethane (PU) | |
No | |
Unisex | |
Uvex 1 | |
No | |
SRC |
Microvelour | |
Steel | |
EN ISO 20345:2011 | |
Wide width | |
Yes | |
S3 |