Learn More
Cytiva Buffer Tray for AKTA™ start
Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
ÄKTA Start
Unit Size:
Each
Description
Buffer bottle tray for use in the AKTA start automated protein purification system.
Specifications
Specifications
| Description | Buffer Bottle Tray |
| For Use With (Equipment) | ÄKTA Start |