Learn More
Buffer solution, pH 12.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
408.60 NOK - 1216.80 NOK
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11316099
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042425.AP
500 mL
408.60 NOK
500mL
View alternative products
Save Up to 
»
In Stock
to see stock.
11314598
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042425.K2
1 L
655.20 NOK
1 L
View alternative products
Save Up to 
»
In Stock
to see stock.
11304598
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042425.K7
4 L
1216.80 NOK
4 L
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
Description
Buffer solution (pH 12.00) is used as a means of keeping the pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.