missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN1A1/Butyrophilin Antibody (2151B) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB84671JF585
This item is not returnable.
View return policy
Description
BTN1A1/Butyrophilin Monoclonal antibody specifically detects BTN1A1/Butyrophilin in Human samples. It is validated for Flow Cytometry,Immunocytochemistry
Specifications
| BTN1A1/Butyrophilin | |
| Monoclonal | |
| Janelia Fluor 585 | |
| BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1 | |
| Mouse myeloma cell line NS0-derived recombinant human BTN1A1/Butyrophilin, Ala27-Arg242, Accession # Q13410 | |
| RUO | |
| 696 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunocytochemistry | |
| 2151B | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction