Learn More
Paragon Scientific™ BS/IP/SL(S) Viscometer for Transparent Liquids (UKAS ISO 17025 certified)
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
Brand: Paragon Scientific™ SHORT07UKC
Additional Details : Weight : 0.40000kg
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
- Supplied with full UKAS ISO 17025 calibration
Specifications
200 to 1000 cSt | |
1 | |
ASTM D 445, D 446 and ISO 3104, 3105 | |
7 |
Yes | |
D 445, D 446 | |
UKAS ISO 17025 calibration |
8 week lead-time