missing translation for 'onlineSavingsMsg'
Learn More

Brucine, 99%, anhydrous, Thermo Scientific Chemicals

Catalog Number p-3735450
Click to view available options
Quantity:
5 g
50 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
357-57-3
C23H26N2O4
394.46
MFCD00005942
RRKTZKIUPZVBMF-VLTSECPOSA-N
brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy
51413923
(4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC
This item is not returnable. View return policy
357-57-3
C23H26N2O4
394.46
MFCD00005942
RRKTZKIUPZVBMF-VLTSECPOSA-N
brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy
51413923
(4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 174.0°C to 179.0°C
Color White to Beige
Loss on Drying 1% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 27, IV, 7876
Merck Index 15, 1464
Specific Rotation Condition −123° (20°C c=2.5,CCl3)
Specific Rotation − 123.00
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, chloroform and benzene, slightly soluble in ether, glycerol and, ethyl acetate
Formula Weight 394.46
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Brucine, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'