missing translation for 'onlineSavingsMsg'
Learn More

Bromoferrocene, 95%, Thermo Scientific Chemicals

Catalog Number p-7028674
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1273-73-0
C10H9BrFe
264.93
MFCD03093913
Bromoferrocene
[Fe].c1cccc1.Brc1cccc1
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1273-73-0
C10H9BrFe
264.93
MFCD03093913
Bromoferrocene
[Fe].c1cccc1.Brc1cccc1

Bromoferrocene is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoferrocene is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 31°C to 33°C
Quantity 1 g
Formula Weight 264.93
Percent Purity 95%
Chemical Name or Material Bromoferrocene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'