missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclopropane, 99%, Thermo Scientific Chemicals

Catalog Number p-7022291
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
4333-56-6
C3H5Br
120.977
MFCD00001271
LKXYJYDRLBPHRS-UHFFFAOYSA-N
cyclopropyl bromide, cyclopropane, bromo, cyclopropylbromide, bromo-cyclopropan, bromo-cyclopropane, cyclopropylbrornide, bromanylcyclopropane, pubchem14417, bromocyclopropane, acmc-1aii5
78037
bromocyclopropane
C1CC1Br
This item is not returnable. View return policy
4333-56-6
C3H5Br
120.977
MFCD00001271
LKXYJYDRLBPHRS-UHFFFAOYSA-N
cyclopropyl bromide, cyclopropane, bromo, cyclopropylbromide, bromo-cyclopropan, bromo-cyclopropane, cyclopropylbrornide, bromanylcyclopropane, pubchem14417, bromocyclopropane, acmc-1aii5
78037
bromocyclopropane
C1CC1Br

Bromo Cyclopropane is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It is also used in the production of cyclopropyl boric acid. It is used as an intermediate in organic synethesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromo Cyclopropane is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It is also used in the production of cyclopropyl boric acid. It is used as an intermediate in organic synethesis.

Solubility
It is soluble in chloroform, and methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY
Density 1.515
Boiling Point 68°C to 70°C
Flash Point −6°C (21°F)
Odor Recognizable
Refractive Index 1.46
Quantity 10 g
UN Number UN1993
Beilstein 1900287
Solubility Information It is soluble in chloroform,and methanol. Insoluble in water.
Formula Weight 120.98
Percent Purity 99%
Chemical Name or Material Bromocyclopropane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'