Learn More
Bromoacetaldehyde dimethyl acetal, 97%, stabilized, ACROS Organics™
Brand:  Acros Organics 221871000
Additional Details : CAS Number : 7252-83-7 Weight : 0.19300kg Transport : UN number : 2929 Chem class : 6.1 Pack group : II
Packaging Glass bottle
Quantity 100mL
 View more versions of this product
Product Code. 10042840
  • 1216.00 NOK / 100mL
In stock, ready to ship: 2
Chemical Identifiers
7252-83-7
169.02
FUSFWUFSEJXMRQ-UHFFFAOYSA-N
81672
COC(CBr)OC
C4H9BrO2
MFCD00000213
bromoacetaldehyde dimethyl acetal, ethane, 2-bromo-1,1-dimethoxy, 2,2-dimethoxyethyl bromide, dimethylbromoacetal, 1-bromo-2,2-dimethoxyethane, 2,2-dimethoxybromoethane, 2-bromoacetaldehyde dimethyl acetal, bromoacetoaldehyde dimethyl acetal, bromoacetaldehyde dimethylacetal, acetaldehyde, bromo-, dimethyl acetal
2-bromo-1,1-dimethoxyethane
Specifications
Bromoacetaldehyde dimethyl acetal
51°C
1.4450 to 1.4470
148.0°C to 150.0°C
97%
96% min. (GC)
BrCH2CH(OCH3)2
01, 624
Solubility in water: insoluble
COC(CBr)OC
169.02
169.02
1.4300g/mL
Glass bottle
1.43
100mL
7252-83-7
C4H9BrO2
MFCD00000213
bromoacetaldehyde dimethyl acetal, ethane, 2-bromo-1,1-dimethoxy, 2,2-dimethoxyethyl bromide, dimethylbromoacetal, 1-bromo-2,2-dimethoxyethane, 2,2-dimethoxybromoethane, 2-bromoacetaldehyde dimethyl acetal, bromoacetoaldehyde dimethyl acetal, bromoacetaldehyde dimethylacetal, acetaldehyde, bromo-, dimethyl acetal
FUSFWUFSEJXMRQ-UHFFFAOYSA-N
2-bromo-1,1-dimethoxyethane
81672
97%
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Bromoacetaldehyde dimethyl acetal, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
SDS Documents