missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromcresol Purple EU Pharmacopoeia, Fisher Chemical™
Click to view available options
Quantity:
100 mL
Packaging:
Bottle
Specifications
Packaging | Bottle |
Quantity | 100 mL |
Grade | EU Ph Ref: 1012601 |
Chemical Name or Material | Bromocresol purple solution. |
Safety and Handling
Product Identifier
- Bromocresol purple solution. European Pharmacopoeia Ref: 1012701. (RP 038)
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'