missing translation for 'onlineSavingsMsg'
Learn More

BPGM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17173-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847014

  • 1415.13 NOK / 20µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

BPGM belongs to the phosphoglycerate mutase family, BPG-dependent PGAM subfamily. It plays a major role in regulating hemoglobin oxygen affinity as a consequence of controlling 2, 3-BPG concentration. It can also catalyze the reaction of EC 5.4.2.1 (mutase) and EC 3.1.3.13 (phosphatase), but with a reduced activity.
Specifications

Specifications

BPGM
Polyclonal
Unconjugated
BPGM
2,3 bisphosphoglycerate mutase, Bisphosphoglycerate mutase, BPG dependent PGAM, BPGM
Rabbit
Antigen Affinity Chromatography
RUO
12183, 296973, 669
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07738, P15327
BPGM
BPGM Fusion Protein Ag10731
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers