Learn More
Duran™ Borosilicate Desiccator Lid with Knob
Fits desiccator base with corresponding flat flange. Duran™ Borosilicate Lid with Knob provides a vacuum-tight seal.
Brand:  Duran™ 244106102
Additional Details : Weight : 1.24000kg
 View more versions of this product
Product Code. 10007820
  • 745.00 NOK / Each
Estimated Shipment
12-02-2020
Add to basket
Description
Description
  • Clear glass
  • Knob lid
  • Fits all desiccator bases with corresponding flat flange
  • Vacuum-tight
Specifications
Specifications
270mm flange
Duran Borosilicate
all base types
Each
Desiccator Lid
107mm