Learn More
Boron solution for ICP 10g/L H2O matrix, Fisher chemical™
Catalog Number p-7083895
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Concentration 10±0.02g/L
Packaging Bottle
Quantity 100 mL
Type Mass Spectrometry Standard
Analyte or Component Names B
Solvent or Matrix H2O
compliance-icons
Product Identifier
  • Single-element Standard Solution for ICP. Boron (B) 10000mg/l in water
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .