missing translation for 'onlineSavingsMsg'
Learn More

Boron, plasma standard solution, Specpure™ B 10,000μg/mL, Thermo Scientific Chemicals

Catalog Number p-7049425
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
MFCD00134034
This item is not returnable. View return policy
MFCD00134034

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration H3BO3 in 1% NH4OH
Quantity 50 mL
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Boron plasma standard solution, B 10000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'