Learn More
Bohlender™ BOLA™ Filtering Sheets
Filtering sheets made of microporous PTFE.
Brand:  Bohlender™ N1617-30
Additional Details : Weight : 0.17700kg
 View more versions of this product
Product Code. 17107524
  • 432.00 NOK / Each
Estimated Shipment: 10-11-2023
to see stock.
Add to basket
Description
Description
  • Made of microporous PTFE
  • Suitable for cutting and blanking
  • Reduction of stability and mechanical load capacity has to be observed when processing materials with larger pore sizes
Specifications
Specifications
PTFE
For Cutting and Blanking
0.2 mm
12 in.
6 in.
5 μm
0.008 in.
300 x 150 mm
300 mm
150 mm