Learn More
Lovibond™ BOD Overflow Flask
For use with Lovibond oxygen measurment systems. Biochemical Oxygen Demand (BOD) measurement is used as a basis for the detection of biologically degradable organic matter in water.
Brand: Lovibond™ 418658
Additional Details : Weight : 0.07600kg
Description
- Overflow flask, 21.7 and 244mL
- For accurate measurement of sample volumes
- For BD 600, BD 606 and Oxi700 BOD Oxygen Meters
Specifications
224 mL | |
BD 600, BD 606 and Oxi700 BOD Oxygen Meters | |
BOD Overflow Flask |
BOD Analysis | |
0 to 200mg/L |