Learn More
Uvex BOAR™ Black/Yellow PU 65667 Low Shoe - Width 10
Uvex 1 safety shoes protect and support the natural movement of the human body by reducing pressure and strain; protection classes S1 and S2
1735.00 NOK
Specifications
Color | Black, Yellow |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17202171
|
Uvex
6566735 |
35 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17212171
|
Uvex
6566736 |
36 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17222171
|
Uvex
6566737 |
37 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17232171
|
Uvex
6566738 |
38 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17242171
|
Uvex
6566739 |
39 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17252171
|
Uvex
6566740 |
40 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17262171
|
Uvex
6566741 |
41 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17272171
|
Uvex
6566742 |
42 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17282171
|
Uvex
6566743 |
43 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17292171
|
Uvex
6566744 |
44 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17202181
|
Uvex
6566745 |
45 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17212181
|
Uvex
6566746 |
46 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17222181
|
Uvex
6566747 |
47 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17232181
|
Uvex
6566748 |
48 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17242181
|
Uvex
6566749 |
49 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17252181
|
Uvex
6566750 |
50 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17262181
|
Uvex
6566751 |
51 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17272181
|
Uvex
6566752 |
52 |
1735.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Uvex 1 Shoe 6566.7 with BOA™ Fit System S2 SRC
- Sporty, multifunctional safety shoe
- Lightweight and flexible
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
- Made with new developed technology for extended wearing comfort
- Uvex Climazone technology for optimum wearer comfort
- Breathable, water-resistant, high-tech microvelour upper material
- Distance mesh lining provides optimum breathability
- Made from synthetic materials, suitable for people allergic to chrome
- BOA™ Fit System Dial for custom comfort that's consistent all day
- Virtually seamless design to eliminate pressure points
- Removable comfort insole, antistatic, with moisture control system and additional shock absorption in heel and at front of foot
- Soft, padded collar and tongue
- Outstanding dual density polyurethane sole unit with best shock absorption characteristics and superb slip resistance
- Free of lacquer wetting inhibitors such as silicones, phthalates, etc.
- Fulfills ESD requirements: resistance to ground Kleiner 35 megohms
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6566.7
- Standard EN ISO 20345:2011 S2 SRC
- Sizes: 35 to 52
Specifications
Black, Yellow | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 1 | |
No | |
SRC |
Microvelour | |
Composite | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S2 |