Learn More
Uvex Blue PU 85329 Boot - Width 12
Uvex 1 safety shoes protect and support the natural movement of the human body by reducing pressure and strain; protection classes S1 and S2
1325.00 NOK
Specifications
Color | Blue |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17204991
|
Uvex
8532935 |
35 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17214991
|
Uvex
8532936 |
36 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17224991
|
Uvex
8532937 |
37 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17234991
|
Uvex
8532938 |
38 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17244991
|
Uvex
8532939 |
39 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17254991
|
Uvex
8532940 |
40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17264991
|
Uvex
8532941 |
41 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17274991
|
Uvex
8532942 |
42 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17284991
|
Uvex
8532943 |
43 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17294991
|
Uvex
8532944 |
44 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17205001
|
Uvex
8532945 |
45 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17215001
|
Uvex
8532946 |
46 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17225001
|
Uvex
8532947 |
47 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17235001
|
Uvex
8532948 |
48 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17245001
|
Uvex
8532949 |
49 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17255001
|
Uvex
8532950 |
50 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17265001
|
Uvex
8532951 |
51 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17275001
|
Uvex
8532952 |
52 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Uvex 1 Boot 8532.9 S1 SRC
- Sporty, multifunctional safety shoe
- Especially lightweight and flexible
- 100% metal-free Uvex Xenova™ toecap
- Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
- Made with new developed last technology for extended wearing comfort
- Uvex Climazone technology ensures optimum wearer comfort
- Breathable, perforated, high-tech microvelour upper material
- Distance mesh lining provides optimum breathability
- Made from synthetic materials, suitable for people allergic to chrome
- Virtually seamless design to eliminate pressure points
- Removable comfort insole, antistatic with moisture control system and shock absorption in heel and at front of foot
- Soft padded collar and tongue
- Outstanding dual density polyurethane sole unit with best shock absorption characteristics and superb slip resistance
- Free of lacquer wetting inhibitors such as silicones, phthalates, etc.
- Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 8532.9
- Standard EN ISO 20345:2011 S1 SRC
- Sizes: 35 to 52
Specifications
Blue | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 1 | |
No | |
SRC |
Microvelour | |
Composite | |
EN ISO 20345:2011 | |
Wide width | |
Yes | |
S1 |