missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Blue Chemical Resistant Gloves, Model CS710

Product Code. 17837361
Click to view available options
Size:
7
8
9
10
11
Unit Size:
6 pairs
This item is not returnable. View return policy
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.

Product Code. 17837361

Brand: SHOWA™ CS71010XL

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.

Prioritize comfort and wearer safety with the CS710 chemical-resistant gloves.

  • A double-dipped nitrile coating offers robust durability, whilst the seamless liner and natural latex-free composition ensure comfort during long periods of wear.
  • Enhanced grip from the foamed nitrile palm coating provides extra grip, even in oily or slippery environments.

Specifications

Chlorinated No
Color Blue
Finger Thickness (English) 57.8 mil
Finger Thickness (Metric) 1.47 mm
Latex Free Yes
Material Nitrile, Polyester
Palm Thickness (English) 57.8 mil
Palm Thickness (Metric) 1.47 mm
Size 10
Thickness (English) 57.8 mil
Thickness (Metric) 1.47 mm
EN388 Standard EN388:4122A
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLMNOPT
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Lining Lined
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.