Learn More
Uvex Blue 85319 Low Shoe - Width 12
Uvex 1 safety shoes protect and support the natural movement of the human body by reducing pressure and strain; protection classes S1 and S2
1280.00 NOK
Specifications
Color | Blue |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17284911
|
Uvex
8531935 |
35 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17294911
|
Uvex
8531936 |
36 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17204921
|
Uvex
8531937 |
37 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17214921
|
Uvex
8531938 |
38 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17224921
|
Uvex
8531939 |
39 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17234921
|
Uvex
8531940 |
40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17244921
|
Uvex
8531941 |
41 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17254921
|
Uvex
8531942 |
42 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17264921
|
Uvex
8531943 |
43 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17274921
|
Uvex
8531944 |
44 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17284921
|
Uvex
8531945 |
45 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17294921
|
Uvex
8531946 |
46 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17204931
|
Uvex
8531947 |
47 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17214931
|
Uvex
8531948 |
48 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17224931
|
Uvex
8531949 |
49 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17234931
|
Uvex
8531950 |
50 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17244931
|
Uvex
8531951 |
51 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17254931
|
Uvex
8531952 |
52 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Uvex 1 Shoe 8531.9 S1 SRC
- Sporty, multifunctional safety shoe
- Especially lightweight and flexible
- 100% metal-free Uvex Xenova™ toecap
- Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
- Made with new developed last technology for extended wearing comfort
- Uvex Climazone technology ensures optimum wearer comfort
- Breathable, perforated, high-tech microvelour upper material
- Distance mesh lining provides optimum breathability
- Made from synthetic materials, suitable for people allergic to chrome
- Virtually seamless design to eliminate pressure points
- Removable comfort insole, antistatic with moisture control system and shock absorption in heel and at front of foot
- Soft padded collar and tongue
- Outstanding dual density polyurethane sole unit with best shock absorption characteristics and superb slip resistance
- Free of lacquer wetting inhibitors such as silicones, phthalates, etc.
- Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 8531.9
- Standard EN ISO 20345:2011 S1 SRC
- Sizes: 35 to 52
Specifications
Blue | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 1 | |
No | |
SRC |
Microvelour | |
Composite | |
EN ISO 20345:2011 | |
Wide width | |
Yes | |
S1 |