Learn More
Uvex Black/Orange Rubber/PU 65231 Boot - Width 10
Optimal shock absorption, ergonomic fit, low weight and excellent climate characteristics ensure max performance at work and redefine limits for safety shoes; range in protection classes S3, S2 and S1 P
1915.00 NOK
Specifications
Color | Black, Orange |
---|---|
Material | Leather |
Sole Material | Dual-density polyurethane rubber (PU/RU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17211201
|
Uvex
6523135 |
35 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17221201
|
Uvex
6523136 |
36 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17231201
|
Uvex
6523137 |
37 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17241201
|
Uvex
6523138 |
38 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17251201
|
Uvex
6523139 |
39 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17261201
|
Uvex
6523140 |
40 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17271201
|
Uvex
6523141 |
41 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17281201
|
Uvex
6523142 |
42 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17291201
|
Uvex
6523143 |
43 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17201211
|
Uvex
6523144 |
44 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17211211
|
Uvex
6523145 |
45 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17221211
|
Uvex
6523146 |
46 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17231211
|
Uvex
6523147 |
47 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17241211
|
Uvex
6523148 |
48 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17251211
|
Uvex
6523149 |
49 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17261211
|
Uvex
6523150 |
50 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17271211
|
Uvex
6523151 |
51 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17281211
|
Uvex
6523152 |
52 |
1915.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 VIBRAM Lace-Up Boot 6523.1 S3 HI HRO SRC
- Sporty, multifunctional safety boots
- Extremely light and flexible
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side and ankle twist stability, no thermal conductivity and anti-magnetic properties
- Metal-free, penetration-resistance insole in accordance with latest standard, which does not impact shoe flexibility
- Made with new high-performance technology for exceptional wearer comfort
- Uvex Climazone technology for optimum comfort
- Breathable, water-resistant leather upper material
- Distance mesh lining provides optimum breathability
- Reflecting elements for more safety
- Virtually seam-free construction to eliminate pressure points
- Removable comfort insole, antistatic, with moisture control system and additional shock absorption in heel and at front of foot
- Soft, padded collar and tongue, dust tongue
- Injection-molded polyurethane scuffcap to protect upper leather on toe cap and significantly extend resistance time
- Meets ESD requirements: resistance to ground Kleiner of 35 megohms
- Lightweight two-layer insole with excellent shock absorption characteristics and outstanding slip resistance
- Multilayer sole system comprising PU midsole and highly slip resistant rubber outersole from VIBRAM
- Optimized ankle protection to ensure safety when working on ladders
- Rugged, self-cleaning sole ridges ensure max grip for demanding applications on uneven or dirty floors (SRC labeling)
- Withstands brief exposure to heat approx. 300°C
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6523.1
- Standard EN ISO 20345:2011 S3 HI HRO SRC
- Sizes: 35 to 52
Specifications
Black, Orange | |
Dual-density polyurethane rubber (PU/RU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Leather | |
Composite | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S3 |