missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BLACK HRP Staining Kit (Colorimetric)
Assay Kit (Colorimetric)
Brand: Novus Biologicals™ NBP3-12183
This item is not returnable.
View return policy
Description

Specifications
30 mL | |
BLACK HRP | |
Store at 4°C. | |
Immunohistochemistry (IHC), Immunohistochemistry (Paraffin), ISH |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction