missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(cyclopentadienyl)dimethylzirconium(IV), 98+%

Catalog Number p-7029815
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
12636-72-5
C12H20Zr
255.516
MFCD01073786
KZBPGWFJSPGCMX-UHFFFAOYSA-N
Dimethylbis(cyclopentadienyl)zirconium(IV); Zirconocene dimethyl
131674679
cyclopenta-1,3-diene;methane;zirconium
C.C.C1C=CC=C1.C1C=CC=C1.[Zr]
This item is not returnable. View return policy
12636-72-5
C12H20Zr
255.516
MFCD01073786
KZBPGWFJSPGCMX-UHFFFAOYSA-N
Dimethylbis(cyclopentadienyl)zirconium(IV); Zirconocene dimethyl
131674679
cyclopenta-1,3-diene;methane;zirconium
C.C.C1C=CC=C1.C1C=CC=C1.[Zr]
Melting Point ∽170°C (decomposition)
Quantity 1 g
Formula Weight 251.48
Percent Purity ≥98%
Chemical Name or Material Bis(cyclopentadienyl)dimethylzirconium(IV)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'