missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), 98%, Thermo Scientific Chemicals

Catalog Number p-7051107
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
14481-08-4
C22H38NiO4
425.24
MFCD00192348
FPQQRRXEOLXSCJ-UHFFFAOYSA-N
bis 2,2,6,6-tetramethyl-3,5-heptanedionate nickel ii, bis 2,2,6,6-tetramethyl-3,5-heptanedionato nickel ii , min-ni ni tmhd 2
131675868
(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;nickel
[Ni++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
This item is not returnable. View return policy
14481-08-4
C22H38NiO4
425.24
MFCD00192348
FPQQRRXEOLXSCJ-UHFFFAOYSA-N
bis 2,2,6,6-tetramethyl-3,5-heptanedionate nickel ii, bis 2,2,6,6-tetramethyl-3,5-heptanedionato nickel ii , min-ni ni tmhd 2
131675868
(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;nickel
[Ni++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C

NMR shift reagent, nickel oxide precursor

NMR shift reagent, nickel oxide precursor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 223°C to 225°C
Boiling Point 90°C to 110°C (0.1 mmHg)(sublimation)
Quantity 1 g
Formula Weight 425.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'