missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Biotinylated Human LAG-3 / CD223 Protein, Fc,Avitag™
MALS verified
5341.70 NOK - 23503.49 NOK
Specifications
Accession Number | NP_002277.4 |
---|---|
Molecular Weight (g/mol) | 75.0 kDa |
Source | Biotinylated Human LAG-3, Fc,Avitag (LA3-H82Fb) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession # P18627-1). |
Immunogen | Leu 23 - Leu 450 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16447829
|
ACROBiosystems
LA3-H82FB-25UG |
25 μg |
5341.70 NOK
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16437829
|
ACROBiosystems
LA3-H82FB-200UG |
200 μg |
23503.49 NOK
200µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Lymphocyte activation gene 3 protein (LAG3) is also known as CD antigen CD223 and protein FDC, which belongs to immunoglobulin (Ig) superfamily and contains 4 extracellular Ig-like domains. The LAG3 gene contains 8 exons. The sequence data, exon/intron organization, and chromosomal localization all indicate a close relationship of LAG3 to CD4. LAG3 /CD223 involved in lymphocyte activation. LAG3 /CD223 binds to HLA class-II antigens.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 75.0 kDa. The protein migrates as 90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_002277.4 | |
Biotinylated Human LAG-3, Fc,Avitag (LA3-H82Fb) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession # P18627-1). | |
-20°C | |
LAG-3 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
75.0 kDa | |
Leu 23 - Leu 450 | |
LAG3,CD223,FDC | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |