Learn More
ACROBiosystems Biotinylated Human IL-23 R Protein, Fc, Avitag™
Biotinylated Protein
5297.56 NOK - 23309.24 NOK
Specifications
Accession Number | NP_653302.2 |
---|---|
Molecular Weight (g/mol) | 66.3 kDa |
Source | Biotinylated Human IL-23 R, Fc,Avitag (ILR-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Gly 355 (Accession # Q5VWK5-1). |
Immunogen | Gly 24 - Gly 355 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16447779
|
ACROBiosystems
ILR-H82F3-25UG |
25 μg |
5297.56 NOK
25µg |
Estimated Shipment: 10-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16437779
|
ACROBiosystems
ILR-H82F3-200UG |
200 μg |
23309.24 NOK
200µg |
Estimated Shipment: 10-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Interleukin 23 receptor (IL-23R) is a type I cytokine receptor, and IL-23R pairs with the receptor molecule IL12RB1/IL12Rbeta1, and both are required for IL23A signaling. Also, IL-23R associates constitutively with Janus kinase 2 (JAK2), and binds to transcription activator STAT3 in a ligand-dependent manner. Furthermore, IL-23R mediates T-cells, NK cells and possibly certain macrophage/myeloid cells stimulation probably through activation of the Jak-Stat signaling cascade. As for IL-23, it may be responsible for autoimmune inflammatory diseases and be important for tumorigenesis.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 66.3 kDa. The protein migrates as 95-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_653302.2 | |
Biotinylated Human IL-23 R, Fc,Avitag (ILR-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Gly 355 (Accession # Q5VWK5-1). | |
-20°C | |
IL-23 R | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
66.3 kDa | |
Gly 24 - Gly 355 | |
IL-23 R,IL-23 Receptor | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |