Learn More
ACROBiosystems Biotinylated Human CD24 Protein, His, Avitag™
Biotinylated Protein
Brand: ACROBiosystems CD4-H82E9-25ug
Additional Details : Weight : 0.00010kg
Description
CD24 may have a pivotal role in cell differentiation of different cell types. Signaling could be triggered by the binding of a lectin-like ligand to the CD24 carbohydrates, and transduced by the release of second messengers derived from the GPI-anchor. Modulates B-cell activation responses. Promotes AG-dependent proliferation of B-cells, and prevents their terminal differentiation into antibody-forming cells. In association with SIGLEC10 may be involved in the selective suppression of the immune response to danger-associated molecular patterns (DAMPs) such as HMGB1, HSP70 and HSP90. Plays a role in the control of autoimmunity.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 6.7 kDa. The protein migrates as 18-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_001278666.1 | |
25 μg | |
Ser 27 - Gly 59 | |
CD24,CD24A | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
6.7 kDa | |
Biotinylated Human CD24, His,Avitag (CD4-H82E9) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession # P25063-1). | |
-20°C | |
CD24 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |