missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human APRIL / TNFSF13 Protein, Fc, Avitag™

Biotinylated Protein

5341.70 NOK - 23503.49 NOK

Specifications

Accession Number NP_003799.1
Molecular Weight (g/mol) 44.6 kDa
Source Biotinylated Human APRIL, Fc,Avitag (APL-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ala 105 - Leu 250 (Accession # O75888-1).
Immunogen Ala 105 - Leu 250
Storage Requirements -20°C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16437169
View Documents Promotion Details
ACROBiosystems
APL-H82F5-200UG
200 μg
23503.49 NOK
200µg
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16447169
View Documents Promotion Details
ACROBiosystems
APL-H82F5-25UG
25 μg
5341.70 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

APRIL(a proliferation-inducing ligand) is also known as Tumor necrosis factor ligand superfamily member 13, TALL-2, TRDL-1, CD256, TNFFSF 13, cytokine that binds to TNFRSF13B/TACI and to TNFRSF17/BCMA. APRIL is a cytokine of the tumor necrosis factor family associated mainly with hematologic malignancies. The closely related TNF family ligands B cell activation factor (BAFF) and a proliferation-inducing ligand (APRIL) serve in the generation and maintenance of mature B-lymphocytes. Both BAFF and APRIL assemble as homotrimers that bind and activate several receptors that they partially share. BAFF-APRIL heteromers of different stoichiometries have distinct receptor-binding properties and activities. In addition, expression of APRIL was regulated by miR-145 in GC cells.

This protein carries a human IgG1 Fc tag at the N-terminus, followed by an Avi tag. The protein has a calculated MW of 44.6 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

NP_003799.1
Biotinylated Human APRIL, Fc,Avitag (APL-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ala 105 - Leu 250 (Accession # O75888-1).
-20°C
APRIL
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
MABSol™ Biotin Labeled Proteins
44.6 kDa
Ala 105 - Leu 250
APRIL,TNFSF13,TALL-2,TRDL-1,CD256,TALL2,ZTNF2
Precision Avi
Fc Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Videos
SDS
Documents

Documents

Product Certifications