Learn More
ACROBiosystems Biotinylated Cynomolgus PD-1 / PDCD1 Protein, His, Avitag™
Biotinylated Protein
5341.70 NOK - 23503.49 NOK
Specifications
Accession Number | NP_001271065 |
---|---|
Molecular Weight (g/mol) | 19.5 kDa |
Source | Biotinylated Cynomolgus PD-1, His,Avitag (PD1-C82E6) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ3). |
Immunogen | Leu 25 - Gln 167 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16437949
|
ACROBiosystems
PD1-C82E6-200UG |
200 μg |
23503.49 NOK
200µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16447949
|
ACROBiosystems
PD1-C82E6-25UG |
25 μg |
5341.70 NOK
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Programmed cell death protein 1 (PD-1) is also known as CD279 and PDCD1, is a type I membrane protein and is a member of the extended CD28/CTLA-4 family of T cell regulators. PDCD1 is expressed on the surface of activated T cells, B cells, macrophages, myeloid cells and a subset of thymocytes. PD-1 has two ligands, PD-L1 and PD-L2, which are members of the B7 family. PD-L1 is expressed on almost all murine tumor cell lines, including PA1 myeloma, P815 mastocytoma, and B16 melanoma upon treatment with IFN-γ. PD-L2 expression is more restricted and is expressed mainly by DCs and a few tumor lines. PD1 inhibits the T-cell proliferation and production of related cytokines including IL-1, IL-4, IL-10 and IFN-γ by suppressing the activation and transduction of PI3K/AKT pathway. In addition, coligation of PD1 inhibits BCR-mediating signal by dephosphorylating key signal transducer. In vitro, treatment of anti-CD3 stimulated T cells with PD-L1-Ig results in reduced T cell proliferation and IFN-γ secretion. Monoclonal antibodies targeting PD-1 that boost the immune system are being developed for the treatment of cancer.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 19.5 kDa. The protein migrates as 30-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_001271065 | |
Biotinylated Cynomolgus PD-1, His,Avitag (PD1-C82E6) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ3). | |
-20°C | |
PD-1 | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
19.5 kDa | |
Leu 25 - Gln 167 | |
PDCD1,PD1,CD279,SLEB2 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |