Learn More
BioTek™ Microplate Washer Flat Magnets
For insertion into BioTek washer biomagnetic separation carriers
Brand:  BioTek™ 7103017
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12544197
  • 20359.00 NOK / Each
Estimated Shipment: 24-11-2021
Description
Description
In flat-bottom well, beads pulled to band across well bottom. In round-bottom well, beads pulled to button at well bottom.
Order Info
For use with: 405™ LS , 405™ Touch, EL406, ELx405, ELx50 washers
Specifications
Specifications
405 ls, 405 touch, el406, elx405, elx50 microplate washers
384-Well Flat Magnet