Learn More
BioTek™ 405™ LS Microplate Washers For 96 Well Microplate Washing
Fifth generation washer automates basic 96-well dispensing and aspiration
Brand:  BioTek™ 405LSR
Additional Details : Weight : 13.50000kg
 View more versions of this product
Product Code. 12573557
  • 84472.00 NOK / Each
Estimated Shipment: 16-09-2021
Description
Description
  • For easy processing of cell based assays, ELISA and multiplex assays
  • Programmable in minutes and seconds, up to 60 minutes (variable intensities)
  • Waste and vacuum sensing, fluid and flow detection
  • Aerosol cover and overflow protection
Hardware
  • Accommodates 96 well microplates
  • 2 x 24 character backlit LCD display and membrane keypad user interface
  • Automated internal four buffer switching
  • Internal positive displacement pump eliminates use of troublesome pressure delivery systems and allows use of any size dispense reservoir
  • Built-in priming trough allows automatic manifold priming and rinsing during both operation and maintenance modes
  • Quick change manifold design
  • BioStack and third party automation compatible
  • All washers biomagnetic separation ready (accessory magnets sold separately)
  • All washers vacuum filtration ready (accessory filtration modules sold separately)
Software
Quick menu allows fluid paths to be easily primed, wash process started and manifold cleaned
Predefined protocols included for out-of-box washing
Fully customizable protocol parameters for individualized wash requirements (gentle cell washing to vigorous wash for high binding assays)
AutoPrime and auto maintenance routines
  • Run protocols created onboard or downloaded from LHC Software
Certifications
CE Marked
Specifications
Specifications
50/60Hz
100 to 240V AC
15° to 30°C
100 to 240V AC 50/60 Hz ≤8A
Washer for 96-well microplate
10 to 85%, non condensing
3 aspirate/dispense cycles, 300μL/well, 96-tube manifold, final aspirate <30seconds