Learn More
Mettler Toledo™ Bioclean™ Filter Pipette Tips
An individually sealed refill rack for LTS™ LiteTouch™ tip racks.
Brand: Mettler Toledo™ 17002428
Additional Details : Weight : 0.63000kg
Description
- Shell and rack made from recycled materials
- 75% less waste than conventional packaging
- Hydrophobic filter forms a solid barrier against aerosols and liquids
- No contamination, no additives, no flakes, just pure protection
- 200μL max. volume
- Pre-sterilized
- 960 tips in 10 rack refills
Specifications
200 μL | |
Sterile | |
960 tips in 10 refills | |
LTS™ LiteTouch™ Tip Racks |
Filtered | |
LiteTouch™ System LTS™ | |
Pipet tip rack refill | |
Bioclean |