missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine tert-butyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog Number p-7055701
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
58620-93-2
C7H16ClNO2
181.66
MFCD00058258
DOMTZTVJNZKUNX-UHFFFAOYSA-N
tert-butyl 3-aminopropanoate hydrochloride, h-beta-ala-otbu.hcl, beta-alanine tert-butyl ester hydrochloride, h-beta-ala-otbu hcl, beta-alanine t-butyl ester hydrochloride, 3-amino-propionic acid tert-butyl ester hydrochloride, h-?-ala-otbu.hcl, h-, a-ala-otbu.hcl, h-beta-ala-otbu hydrochloride, tert-butyl 3-aminopropanoate-hcl
16218890
tert-butyl 3-aminopropanoate;hydrochloride
Cl.CC(C)(C)OC(=O)CCN
This item is not returnable. View return policy
58620-93-2
C7H16ClNO2
181.66
MFCD00058258
DOMTZTVJNZKUNX-UHFFFAOYSA-N
tert-butyl 3-aminopropanoate hydrochloride, h-beta-ala-otbu.hcl, beta-alanine tert-butyl ester hydrochloride, h-beta-ala-otbu hcl, beta-alanine t-butyl ester hydrochloride, 3-amino-propionic acid tert-butyl ester hydrochloride, h-?-ala-otbu.hcl, h-, a-ala-otbu.hcl, h-beta-ala-otbu hydrochloride, tert-butyl 3-aminopropanoate-hcl
16218890
tert-butyl 3-aminopropanoate;hydrochloride
Cl.CC(C)(C)OC(=O)CCN

Useful intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Clear solution (0.3g in 2ml MeOH).

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 167°C to 169°C
Linear Formula H2NCH2CH2COOC(CH3)3·HCl
Beilstein 6660122
Solubility Information Clear solution (0.3g in 2ml MeOH).
Formula Weight 181.66
Percent Purity 95%
Quantity 5 g
Physical Form Solid
Chemical Name or Material beta-Alanine tert-butyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'