missing translation for 'onlineSavingsMsg'
Learn More

Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog Number p-7022299
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
622-78-6
C8H7NS
149.211
MFCD00004819
MDKCFLQDBWCQCV-UHFFFAOYSA-N
benzyl isothiocyanate, isothiocyanatomethyl benzene, benzyl mustard oil, benzylisothiocyanate, benzylsenfoel, tromacaps, tromalyt, urogran, isothiocyanic acid, benzyl ester, benzyl-isothiocyanate
2346
CHEBI:17484
isothiocyanatomethylbenzene
C1=CC=C(C=C1)CN=C=S
This item is not returnable. View return policy
622-78-6
C8H7NS
149.211
MFCD00004819
MDKCFLQDBWCQCV-UHFFFAOYSA-N
benzyl isothiocyanate, isothiocyanatomethyl benzene, benzyl mustard oil, benzylisothiocyanate, benzylsenfoel, tromacaps, tromalyt, urogran, isothiocyanic acid, benzyl ester, benzyl-isothiocyanate
2346
CHEBI:17484
isothiocyanatomethylbenzene
C1=CC=C(C=C1)CN=C=S

Benzyl isothiocyanate is used in a synthesis of 2-phenylpyrimidine-4-thiones from Benzoyl­acetonitrile. Benzyl isothiocyanate has been used in the preparation of S-(N-benzylthiocarbamoyl)-L-glutathione and S-(N-benzylthiocarbamoyl)-L-cysteine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl isothiocyanate is used in a synthesis of 2-phenylpyrimidine-4-thiones from Benzoyl­acetonitrile. Benzyl isothiocyanate has been used in the preparation of S-(N-benzylthiocarbamoyl)-L-glutathione and S-(N-benzylthiocarbamoyl)-L-cysteine.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Density 1.125
Boiling Point 242°C to 243°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.603
Quantity 5 g
UN Number UN2922
Beilstein 386135
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 149.22
Percent Purity 98%
Chemical Name or Material Benzyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'