missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloromethyl ether, tech. 70%, Thermo Scientific Chemicals

Catalog Number p-7040435
Click to view available options
Quantity:
50 mL
250 mL
This item is not returnable. View return policy
3587-60-8
C8H9ClO
156.609
MFCD00000886
LADPCMZCENPFGV-UHFFFAOYSA-N
benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl
137983
chloromethoxymethylbenzene
C1=CC=C(C=C1)COCCl
This item is not returnable. View return policy
3587-60-8
C8H9ClO
156.609
MFCD00000886
LADPCMZCENPFGV-UHFFFAOYSA-N
benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl
137983
chloromethoxymethylbenzene
C1=CC=C(C=C1)COCCl

Benzyl chloromethyl ether is employed in the introduction of benzoyloxy- and hydroxymethyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl chloromethyl ether is employed in the introduction of benzoyloxy- and hydroxymethyl groups.

Solubility
Soluble in water [2.7 g/L (25 C)]

Notes
Store at 2-8°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.126
Boiling Point 102°C
Linear Formula C6H5CH2OCH2Cl
Refractive Index 1.527
Quantity 50 mL
UN Number UN2810
Beilstein 1364034
Solubility Information Soluble in water [2.7g/L (25 C)]
Formula Weight 156.61
Percent Purity 70%
Grade Technical
Chemical Name or Material Benzyl chloromethyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'