Learn More
Benzo[d]isoxazol-3-ol, 96%, Thermo Scientific™
Catalog Number p-4813335
Click to view available options
Quantity:
1 g
5 g
21725-69-9
C7H5NO2
135.12
QLDQYRDCPNBPII-UHFFFAOYSA-N
benzo d isoxazol-3-ol, 1,2-benzisoxazol-3 2h-one, 1,2-benzoxazol-3-ol, 1,2-benzisoxazol-3-ol, 3-hydroxybenzisoxazole, 3-hydroxy-1,2-benzisoxazole, 1,2-benzisoxazolin-3-one, 1,2-benzisoxazol-3-ol 8ci, 2h-1,2-benzoxazol-3-one, benzisoxazolone
210830
1,2-benzoxazol-3-one
C1=CC=C2C(=C1)C(=O)NO2
21725-69-9
C7H5NO2
135.12
QLDQYRDCPNBPII-UHFFFAOYSA-N
benzo d isoxazol-3-ol, 1,2-benzisoxazol-3 2h-one, 1,2-benzoxazol-3-ol, 1,2-benzisoxazol-3-ol, 3-hydroxybenzisoxazole, 3-hydroxy-1,2-benzisoxazole, 1,2-benzisoxazolin-3-one, 1,2-benzisoxazol-3-ol 8ci, 2h-1,2-benzoxazol-3-one, benzisoxazolone
210830
1,2-benzoxazol-3-one
C1=CC=C2C(=C1)C(=O)NO2
Melting Point 137°C to 140°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 135.12
Percent Purity 96%
Chemical Name or Material Benzo[d]isoxazol-3-ol
compliance-icons
Product Identifier
  • Benzo¢d!isoxazol-3-ol, 96%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.