missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzethonium Chloride solution EU Pharmacopoeia, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
Concentration | 0.004M |
Packaging | Bottle |
Quantity | 1000 mL |
Grade | Eu. Ph. Reference |
Chemical Name or Material | Benzethonium chloride solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'