Learn More
Alfa Aesar™ Benzenesulfonyl fluoride, 98%
Brand: Alfa Aesar™ L04689.06
Additional Details : CAS Number : 368-43-4 Weight : 0.00500kg Transport : UN number : 2922 Chem class : 8 Pack group : II
Quantity | 5g |
---|
Chemical Identifiers
368-43-4 | |
160.162 | |
IDIPWEYIBKUDNY-UHFFFAOYSA-N | |
67779 | |
C1=CC=C(C=C1)S(=O)(=O)F |
C6H5FO2S | |
MFCD00007412 | |
phenylsulfonyl fluoride, benzenesulphonyl fluoride, benzenesulfonylfluoride, phenylsulfonylfluoride, benzenesulphonylfluoride, fluorophenylsulfone, pubchem14507, acmc-20alq8, wln: wsfr, benzenesulfonyl fluoride | |
benzenesulfonyl fluoride |
Specifications
Benzenesulfonyl fluoride | |
87°C (188°F) | |
Moisture sensitive | |
5g | |
C6H5FO2S | |
UN2922 | |
phenylsulfonyl fluoride, benzenesulphonyl fluoride, benzenesulfonylfluoride, phenylsulfonylfluoride, benzenesulphonylfluoride, fluorophenylsulfone, pubchem14507, acmc-20alq8, wln: wsfr, benzenesulfonyl fluoride | |
C1=CC=C(C=C1)S(=O)(=O)F | |
160.162 | |
160.17 |
1.334 | |
1.492 | |
203°C to 204°C | |
368-43-4 | |
MFCD00007412 | |
2206835 | |
IDIPWEYIBKUDNY-UHFFFAOYSA-N | |
benzenesulfonyl fluoride | |
67779 | |
98% |