Learn More
Bel-Art™ SP Scienceware™ Vortexer Attachment for Microcentrifuge Tubes
Mix contents of up to 20 tubes at one time. Bel-Art™ SP Scienceware™ Vortexer Attachment for Microcentrifuge Tubes securely holds 1.5ml microcentrifuge tubes in a disk for simultaneous vortexing.
Brand:  Bel-Art™ H37080-0020
Additional Details : Weight : 0.16000kg
Product Code. 10276333
  • 1510.00 NOK / Each
Estimated Shipment: 01-07-2024
Description
  • Tube contents are mixed by holding the shaft against the rubber pad of a vortexer
  • Made of polypropylene with 6-1/2 in. (165mm) Delrin shaft and a 1 in. (25.4mm) diameter ball handle
Specifications
Specifications
Centrifuge Ware
Polypropylene, Delrin
Microcentrifuge tube
16.5 cm