Learn More
Bel-Art™ Quaver™ and Quaverette™ Vibrating Spatulas
Make accurate additions of precious materials as needed and reduce transfer losses.
Brand: Bel-Art™ H36714-0000
Additional Details : Weight : 0.00752kg
Description
- Rotating the toothed thumb wheel vibrates the spatula, giving user control over transfer from spatula to weighing dish
- Make accurate additions of precious materials as needed
- Quaver™ spatula features a flat blade with curved end
Specifications
Flat with Curved End | |
Weighing dish or weighing boat or container | |
7 in. | |
Angled end, toothed thumb wheel |
Quaver™ Vibrating Spatula | |
Accurately transfer small amounts of material, provides precise control of sample transfer from the blade to the weighing dish or weighing boat | |
Stainless Steel | |
18 cm |