Learn More
BCDIN3D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558979
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EPQPWKCYRA AARRLRKLGL HDFDHFHSLA IRGDMPNQIV QILTQDHGME LICCFGNTSW DRSLLLFRAK QTIETHPIPE SLIEKGKEKN RLSFQKQ Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.
BCDIN3D Polyclonal antibody specifically detects BCDIN3D in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
BCDIN3D | |
Polyclonal | |
Unconjugated | |
BCDIN3D | |
BCDIN3 domain containing RNA methyltransfease; BCDIN3 domain containing RNA methyltransferase; BCDIN3 domain-containing protein; BCDIN3D; pre-miRNA 5'-monophosphate methyltransferase; probable methyltransferase BCDIN3D; RNA 5'-monophosphate methyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
144233 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z5W3 | |
BCDIN3D | |
Recombinant protein corresponding to Human BCDIN3D | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only