Learn More
Bayonet 1140 A2B2P3 R D Combination Filter
Brand:  Draeger Safety 6738783
Additional Details : Weight : 0.31500kg
Product Code. 12661925
  • 456.00 NOK / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Specifications
EN 12941/12942
Combination Filter
1 Ea.
1140 A2B2P3 R D
This Filter Can be Used in Respiratory Masks with Rd40 Thread