missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BATF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564192
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TEELKYFTSV LNSHEPLCSV LAASTPSPPE VVYSAHAFHQ PHV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
BATF Polyclonal antibody specifically detects BATF in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
BATF | |
Polyclonal | |
Unconjugated | |
Batf | |
activating transcription factor B; basic leucine zipper ATF-like transcription factor; basic leucine zipper transcription factor, ATF-like; basic leucine zipper transcription factor-like; basic leucine zipper transcriptional factor ATF-like; BATF; B-ATF; BATF1; B-cell activating transcription factor; B-cell-activating transcription factor; SFA2; SFA-2; SF-HT-activated gene 2 protein; si:ch211-153j24.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10538 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16520 | |
Batf | |
Recombinant protein corresponding to Human BATF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only