missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium diphenylamine-4-sulfonate, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Barium diphenylamine-4-sulfonate is used as a pharmaceutical intermediate and in chemical research. It is also used as an analytical dosing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Barium diphenylamine-4-sulfonate is used as a pharmaceutical intermediate and in chemical research. It is also used as an analytical dosing agent.
Solubility
It is moderate soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Incompatible with mineral acids, organic acids, strong oxidizing agents.
Barium diphenylamine-4-sulfonate is used as a pharmaceutical intermediate and in chemical research. It is also used as an analytical dosing agent.
Solubility
It is moderate soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Incompatible with mineral acids, organic acids, strong oxidizing agents.
Specifications
Melting Point | >300°C |
Quantity | 5 g |
UN Number | UN1564 |
Solubility Information | It is moderate soluble in water. |
Formula Weight | 633.9 |
Chemical Name or Material | Barium diphenylamine-4-sulfonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'